Merck
Quantification of benzodiazepines in whole blood and serum.
International journal of legal medicine (2005-10-13)
Franz E Dussy, Cornelia Hamberg, Thomas A Briellmann
ABSTRACT
A high-performance liquid chromatography method for the determination of benzodiazepines and their metabolites in whole blood and serum using mass spectrometry (MS) and photodiode array (PDA) detection is presented. The combination of both detection types can complement each other and provides extensive case relevant data. The limits of quantification (LOQ) with the MS detection lie between 2 and 3 microg/l for the following benzodiazepines or metabolites: 7-amino-flunitrazepam, alprazolam, desalkyl-flurazepam, desmethyl-flunitrazepam, diazepam, flunitrazepam, flurazepam, alpha-hydroxy-midazolam, lorazepam, midazolam, nitrazepam, nordazepam and oxazepam, respectively 5 microg/l for lormetazepam and 6 microg/l for bromazepam. The LOQ of clobazam determined with the PDA detector is 10 microg/l. A convenient approach for determining the measurement uncertainty of the presented method--applicable also for other methods in an accreditation process--is presented. At low concentrations (<10 microg/l), measurement uncertainty was estimated to be about 50%, and at concentrations >180 microg/l, it was estimated to be about 15%. One hundred and twenty-eight case data acquired over 1 year are summarised.
Product Description
Supelco
7-Aminoflunitrazepam solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
7-Aminoflunitrazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®