Merck
Chemical aspects of siderophore mediated iron transport.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2002-10-31)
Hakim Boukhalfa, Alvin L Crumbliss
ABSTRACT
In this mini-review we describe selected aspects of the coordination chemistry relevant to siderophore mediated iron transport and bioavailability. Specific emphasis is placed on a discussion of in vitro kinetic and thermodynamic data that are relevant to elucidating possible in vivo mechanisms for environmental iron acquisition by microbial cells.
Product Description
Millipore
Ferrioxamine E from Streptomyces antibioticus, suitable for microbiology, ≥95%