Merck
Dermal pharmacokinetics of the insecticide furathiocarb in rats.
Pest management science (2002-02-13)
Kwang-Hyeon Liu, Ha-Jung Sung, Hae-Keun Lee, Byung-Hun Song, Yang-Bin Ihm, Kyun Kim, Hye-Suk Lee, Jeong-Han Kim
ABSTRACT
The pharmacokinetics of furathiocarb were studied in vivo in male Sprague-Dawley rats following dermal treatment. HPLC and post-column derivatization were used for the analysis of furathiocarb and its metabolites (carbofuran, 3-hydroxycarbofuran and 3-ketocarbofuran). Carbofuran and 3-hydroxycarbofuran were detected in plasma and urine rather than furathiocarb. 3-Ketocarbofuran, another potential metabolite, was not observed in any sample. The concentration of carbofuran was higher than that of 3-hydroxycarbofuran in plasma, but the reverse was the case in urine. The corresponding area under the plasma concentration-time curve, Tmax, and Cmax values of carbofuran and 3-hydroxycarbofuran for 1500 mg kg-1 doses were 2.4-8.0 mg equiv hml-1, 12 h and 0.1-0.4 mg equiv ml-1, respectively. T1/2 was calculated only for 3-hydroxycarbofuran (28 h). Two metabolites were excreted in a dose-dependent manner without saturation.
Product Description
Supelco
Furathiocarb, PESTANAL®, analytical standard
Supelco
Carbofuran-3-hydroxy, PESTANAL®, analytical standard