Merck
  • Asymmetric strecker synthesis of alpha-amino acids via a crystallization-induced asymmetric transformation using (R)-phenylglycine amide as chiral auxiliary.
Asymmetric strecker synthesis of alpha-amino acids via a crystallization-induced asymmetric transformation using (R)-phenylglycine amide as chiral auxiliary.
Organic letters (2001-05-12)
W H Boesten, J P Seerden, B de Lange, H J Dielemans, H L Elsenberg, B Kaptein, H M Moody, R M Kellogg, Q B Broxterman
ABSTRACT
[reaction: see text]. Diastereoselective Strecker reactions based on (R)-phenylglycine amide as chiral auxiliary are reported. The Strecker reaction is accompanied by an in situ crystallization-induced asymmetric transformation, whereby one diastereomer selectively precipitates and can be isolated in 76-93% yield and dr > 99/1. The diastereomerically pure alpha-amino nitrile obtained from pivaldehyde (R1 = t-Bu, R2 = H) was converted in three steps to (S)-tert-leucine in 73% yield and >98% ee.
Sigma-Aldrich
N-Phenylglycine, 97%