Merck
  • Long-lasting cholecystokinin(2) receptor blockade after a single subcutaneous injection of YF476 or YM022.
Long-lasting cholecystokinin(2) receptor blockade after a single subcutaneous injection of YF476 or YM022.
British journal of pharmacology (2000-05-24)
M Kitano, P Norlén, X Q Ding, S Nakamura, R Håkanson
ABSTRACT
Histamine-forming ECL cells in the rat stomach operate under the control of gastrin. They represent a convenient target for studying cholecystokinin-B/gastrin (CCK(2)) receptor antagonists in vivo. We examined the effectiveness and duration of action of two CCK(2) antagonists, YM022 and YF476, with respect to their effect on ECL-cell histidine decarboxylase (HDC) activity in the rat. Oral administration of subcutaneous deposition of YF476 or YM022 reduced the HDC activity. The maximum/near-maximum dose for both drugs and for both modes of administration was 300 micromol kg(-1) (effects measured 24 h after dose). At this dose and time the serum concentration of YF476 was 20 - 40 nmol l(-1). The dose 300 micromol kg(-1) was used in all subsequent studies. A single subcutaneous injection of YF476 inhibited the HDC activity for 8 weeks. The circulating concentration of YF476 remained high for the same period of time (>/=15 nmol l(-1)). Subcutaneous YM022 suppressed the HDC activity for 4 weeks. A single oral dose of YF476 or YM022 inhibited the HDC activity for 2 - 3 days. Chronic gastric fistula rats were used to study the effect of subcutaneous YF476 on gastrin-stimulated acid secretion. A single injection of YF476 prevented gastrin from causing an acid response for at least 4 weeks (the longest time studied). We conclude that a single subcutaneous injection of 300 micromol kg(-1) YF476 causes blockade of CCK(2) receptors in the stomach of the rat for 8 weeks thus providing a convenient method for studies of the consequences of long-term CCK(2) receptor inhibition.
Sigma-Aldrich
YM 022, ≥98% (HPLC)