Merck
  • Salinomycin inhibits prostate cancer growth and migration via induction of oxidative stress.
Salinomycin inhibits prostate cancer growth and migration via induction of oxidative stress.
British journal of cancer (2012-01-05)
K Ketola, M Hilvo, T Hyötyläinen, A Vuoristo, A-L Ruskeepää, M Orešič, O Kallioniemi, K Iljin
ABSTRACT
We have shown that a sodium ionophore monensin inhibits prostate cancer cell growth. A structurally related compound to monensin, salinomycin, was recently identified as a putative cancer stem cell inhibitor. The growth inhibitory potential of salinomycin was studied in a panel of prostate cells. To get insights into the mechanism of action, a variety of assays such as gene expression and steroid profiling were performed in salinomycin-exposed prostate cancer cells. Salinomycin inhibited the growth of prostate cancer cells, but did not affect non-malignant prostate epithelial cells. Salinomycin impacted on prostate cancer stem cell functions as evidenced by reduced aldehyde dehydrogenase activity and the fraction of CD44(+) cells. Moreover, salinomycin reduced the expression of MYC, AR and ERG, induced oxidative stress as well as inhibited nuclear factor-κB activity and cell migration. Furthermore, profiling steroid metabolites revealed increased levels of oxidative stress-inducing steroids 7-ketocholesterol and aldosterone and decreased levels of antioxidative steroids progesterone and pregnenolone in salinomycin-exposed prostate cancer cells. Our results indicate that salinomycin inhibits prostate cancer cell growth and migration by reducing the expression of key prostate cancer oncogenes, inducing oxidative stress, decreasing the antioxidative capacity and cancer stem cell fraction.
Product Description
Niclosamide (anhydrous), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Niclosamide