Merck
  • Induction of CYP3A4 and MDR1 gene expression by baicalin, baicalein, chlorogenic acid, and ginsenoside Rf through constitutive androstane receptor- and pregnane X receptor-mediated pathways.
Induction of CYP3A4 and MDR1 gene expression by baicalin, baicalein, chlorogenic acid, and ginsenoside Rf through constitutive androstane receptor- and pregnane X receptor-mediated pathways.
European journal of pharmacology (2010-06-29)
Yue Li, Qi Wang, Xiaomin Yao, Yan Li
ABSTRACT
The herbal products baicalin, baicalein, chlorogenic acid, and ginsenoside Rf have multiple pharmacological effects and are extensively used in alternative and/or complementary therapies. The present study investigated whether baicalin, baicalein, chlorogenic acid, and ginsenoside Rf induced the expression of the cytochrome P450 3A4 (CYP3A4) and multi-drug resistance 1 (MDR1) genes through the pregnane X receptor and constitutive androstane receptor pathways. Real time PCR, western blotting, and a luminescent assay were used to assess the induction of gene expression and activity of CYP3A4 and MDR1 by the test compounds. The interactions of baicalein/chlorogenic acid/ginsenoside Rf with constitutive androstane receptor and pregnane X receptor were evaluated using luciferase reporter and gel shift assays. Baicalein induced the expression of CYP3A4 and MDR1 mRNA by activating pregnane X receptor and constitutive androstane receptor. Chlorogenic acid and ginsenoside Rf showed a relatively weak effect on CYP3A4 promoter activation only in HepG2 cells cotransfected with constitutive androstane receptor and demonstrated no effects on MDR1 via either the constitutive androstane receptor or pregnane X receptor pathway. Baicalin had no effect on either CYP3A4 or MDR1 gene expression. In conclusion, baicalein has the potential to up-regulate CYP3A4 and MDR1 through the direct activation of the constitutive androstane receptor and pregnane X receptor pathways. Chlorogenic acid and ginsenoside Rf only induced constitutive androstane receptor-mediated CYP3A4 expression.
Product Description
Ginsenoside Rf, primary reference standard
Supelco
Ginsenoside Rf, analytical standard