MilliporeSigma
  • Effects of aldicarb and propoxur on cytotoxicity and lipid peroxidation in CHO-K1 cells.
Effects of aldicarb and propoxur on cytotoxicity and lipid peroxidation in CHO-K1 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-03-30)
E Maran, M Fernández-Franzón, G Font, M J Ruiz
ABSTRACT
Cytotoxic effects of aldicarb, its sulfone and sulfoxide, and propoxur, lipid peroxidation and antioxidant parameters in Chinese Hamster Ovary (CHO-K1) cells were determined. D,L-buthionine-(S,R)-sulfoximine (BSO) was assayed to determine the role of GSH in the protection against carbamate cytotoxicity. Pre-treatment with 60 microM BSO, induced a significant decrease in the glutathione reductase (GR; 64-141%), the glutathione peroxidase (GPx; 10-30%) and the glutathione S-transferase (GST; 59-93%) activities, and its GSH levels (79-85%), while the oxidized glutathione (GSSG) levels significantly increased (64-78%) respect to experiment non-BSO-pretreated. Carbamates BSO pre-treated vs. non-BSO pre-treated showed a significant increase in malondialdehyde (MDA) production (from 13% to 52% vs. 25% to 93%). These data suggest that carbamates could injure CHO-K1 cells via oxidative stress by the increase of MDA production; moreover, BSO enhance the oxidative damage caused by carbamates. However, the glutathione system protects cells from carbamates damage.
Product Description
Supelco
Propoxur, PESTANAL®, analytical standard