Merck
  • Metabolism of fatty acid in yeast: characterisation of beta-oxidation and ultrastructural changes in the genus Sporidiobolus sp. cultivated on ricinoleic acid methyl ester.
Metabolism of fatty acid in yeast: characterisation of beta-oxidation and ultrastructural changes in the genus Sporidiobolus sp. cultivated on ricinoleic acid methyl ester.
FEMS microbiology letters (2005-07-27)
Gilles Feron, Caroline Blin-Perrin, Isabelle Krasniewski, Geneviève Mauvais, Jeannine Lherminier
ABSTRACT
Cell structure modifications and beta-oxidation induction were monitored in two strains of Sporidiobolus, Sp. Ruinenii and Sp. pararoseus after cultivation on ricinoleic acid methyl ester. Ultrastructural observations of the yeast before and after cultivation on fatty acid esters did not reveal major modifications in Sp. ruinenii. Unexpectedly, in Sp. pararoseus a proliferation of the mitochondrion was observed. After induction, Sp. ruinenii principally exhibited an increase in the activities of acyl-CoA oxidase (ACO), hydroxyacyl-CoA deshydrogenase (HAD), thiolase and catalase. In contrast, Sp. pararoseus lacked ACO and catalase activities, but an increase in acyl-CoA deshydrogenase (ACDH) and enoyl-CoA hydratase (ECH) activity was observed. These data suggest that in Sp. ruinenii, beta-oxidation is preferentially localized in the microbody, whereas in Sp. pararoseus it might be localized in the mitochondria.
Sigma-Aldrich
Methyl ricinoleate, ≥99% (GC)
Supelco
Methyl ricinoleate, analytical standard