Merck
  • Copolymers of N-isopropylacrylamide can trigger pH sensitivity to stable liposomes.
Copolymers of N-isopropylacrylamide can trigger pH sensitivity to stable liposomes.
FEBS letters (1998-02-14)
O Meyer, D Papahadjopoulos, J C Leroux
ABSTRACT
Stable liposomes were rendered pH-sensitive by complexation to a polymer that undergoes marked temperature- and pH-dependent water solubility changes. The N-isopropylacrylamide-methacrylic acid copolymer was prepared with or without octadecyl acrylate. At pH below the phase transition of the polymer, egg phosphatidylcholine liposomes quickly released a part of their contents only when associated with the octadecyl aliphatic chain grafted polymer at 37 degrees C. Similarly, sterically stabilized liposomes also quickly released a significant part of the entrapped fluorescent markers at pH 5.5-4.9, values corresponding to those of endosomes/lysosomes. This new pH-sensitive liposome-polymer system may further improve the efficiency of liposomal drug delivery.
Sigma-Aldrich
Poly(N-isopropylacrylamide-co-methacrylic acid), methacrylic acid 10 mol %, Mn 60,000