Merck
  • Regulation of sympathetic presynaptic components in rat left ventricle during ligation of abdominal aorta.
Regulation of sympathetic presynaptic components in rat left ventricle during ligation of abdominal aorta.
The American journal of physiology (1996-10-01)
C Nyquist-Battie, P K Cochran, V R Evans, J M Hitchcock, C Unal
ABSTRACT
To assess the effects of long-term pressure overload on sympathetic presynaptic components in the left ventricle, young adult male rats were subjected to surgical constriction of the suprarenal abdominal aorta. At 4 and 8 wk postsurgery, but not at 1 wk, left ventricular sympathetic activity, measured by the net fractional norepinephrine (NE) decrease after alpha-methyl-p-tyrosine methyl ester administration, was elevated in the aortic-banded rats. However, left ventricular NE was reduced only at 8 wk. Scatchard analysis of saturation binding of [3H]nisoxetine, a specific ligand for NE uptake sites, determined that left ventricular NE transporter sites were also reduced at 8 wk, suggesting a relationship between a reduced number of uptake sites and loss of NE stores. In contrast, aortic constriction did not reduce neuropeptide Y (NPY), tyrosine hydroxylase, dopamine beta-hydroxylase, nervegrowth factor, and low-affinity nerve growth factor receptors at any time point. Thus long-term pressure overload can cause a selective reduction in ventricular NE stores without a reduction in NPY, a colocalized sympathetic neurotransmitter.
Sigma-Aldrich
α-Methyl-DL-tyrosine methyl ester hydrochloride, ≥95% (HPLC)