- Use of endoproteinase Lys-C from Lysobacter enzymogenes in protein sequence analysis.
Use of endoproteinase Lys-C from Lysobacter enzymogenes in protein sequence analysis.
Analytical biochemistry (1983-10-15)
P A Jekel, W J Weijer, J J Beintema
PMID6359954
ABSTRACT
Endoproteinase Lys-C from Lysobacter enzymogenes, which is commercially available, proved to be useful in the determination of primary structures of proteins. The enzyme preferentially cleaves at the carboxyl side of lysine residues.