Merck
  • [Analgesic effect and gastro-intestinal motility inhibitory action of 3-hydroxy-4-methoxy-acetophenone from Cynanchum paniculatum (Bunge) Kitagawa].
[Analgesic effect and gastro-intestinal motility inhibitory action of 3-hydroxy-4-methoxy-acetophenone from Cynanchum paniculatum (Bunge) Kitagawa].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1993-06-01)
F Z Sun, M Cai, F C Lou
ABSTRACT
3-Hydroxy-4-methoxy-acetophenone (HMA) isolated from Cynanchum paniculatum was found to have analgesic effect and inhibitory action on the gastro-intestinal motility. The effect and action were equivalent to that of paeonol, HMA displayed a very low degree of antibacterial activity against Escherichia coli and Shigella flexneri.
Sigma-Aldrich
3′-Hydroxy-4′-methoxyacetophenone, 97%