Merck
  • Modulators of the protein kinase C system influence biliary excretion of cationic drugs.
Modulators of the protein kinase C system influence biliary excretion of cationic drugs.
Hepatology (Baltimore, Md.) (1993-11-01)
H Steen, H Smit, A Nijholt, M Merema, D K Meijer
ABSTRACT
To investigate whether hepatobiliary transport of organic cations is under regulatory control, we studied transport of tri-n-butylmethylammonium in the isolated perfused rat liver and in isolated rat hepatocytes. Transport was investigated in the presence of modulators of the protein kinase C and the cyclic AMP second-messenger system. In the isolated perfused rat liver, it was observed that compounds modulating protein kinase C activity clearly affected the biliary excretion process of the cation tri-n-butylmethylammonium. Phorbol 12-myristate 13-acetate, a compound that directly stimulates protein kinase C, elevated the biliary excretion rate of tri-n-butylmethylammonium in a concentration-dependent manner, reaching a twofold increase at 60 nmol/L of the phorbol ester. The inactive derivative 4 alpha-phorbol 12, 13-didecanoate (60 nmol/L) did not show any effect. Vasopressin (48 nmol/L), a receptor-mediated activator of protein kinase C, stimulated the excretion rate of the cation by about 50%. Staurosporin (1 mumol/L), an inhibitor of protein kinase C, clearly decreased the biliary excretion rate of the cation and also blocked its stimulation by phorbol 12-myristate 13-acetate. Neither phorbol 12-myristate 13-acetate nor vasopressin (at concentrations ranging from 10(-9) to 10(-6) mol/L) affected the initial uptake velocity of tri-n-butylmethylammonium in isolated hepatocytes and isolated perfused livers, whereas staurosporin (1 mumol/L) showed only a modest inhibition of the uptake of the cation. It is inferred that the effect of protein kinase C modulators on hepatobiliary transport of organic cations occurs at the level of carrier-mediated transport in the canalicular membrane.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Tributylmethylammonium chloride solution, 75 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tributylmethylammonium chloride, ≥98.0% (T)