- Blood and urine concentrations of cyclizine by nitrogen-phosphorus gas-liquid chromatography.
Blood and urine concentrations of cyclizine by nitrogen-phosphorus gas-liquid chromatography.
Journal of analytical toxicology (1984-03-01)
D S Griffin, R C Baselt
PMID6716982
ABSTRACT
Gas-liquid chromatography with nitrogen-phosphorus detection was used to analyze blood and urine from a volunteer who ingested 50 mg of cyclizine hydrochloride. A peak blood cyclizine concentration of 69 ng/mL was observed 2 hr after drug administration, and the levels declined thereafter in a biphasic manner, with estimated half-lives of 7 and 24 hr for the early and late phases, respectively. The peak urine cyclizine concentration of 12.5 ng/mL was achieved at 4 hr after administration; only 0.01% of the dose was excreted in the 24 hr urine. Norcyclizine was not observed in blood or urine; however, the detectability of the method for this metabolite is poor relative to the parent drug.