- Effect of corticosterone on adrenal 5-ene-3 beta-hydroxysteroid dehydrogenase in estrogen-treated rats.
Effect of corticosterone on adrenal 5-ene-3 beta-hydroxysteroid dehydrogenase in estrogen-treated rats.
Journal of steroid biochemistry (1983-09-01)
P K Ghosh, N M Biswas
PMID6194376
ABSTRACT
Treatment of adult male rats with estradiol-17 beta for 7 days results in adrenal hyperplasia, increased level of serum ACTH along with reduction in serum level of alpha 2u-globulin and inhibition of adrenal 5-ene-3 beta-hydroxysteroid dehydrogenase (5-ene-3 beta-HSD) activity. Administration of corticosterone in estrogen-treated rats reverses the effects of estrogen while in normal rats corticosterone treatment reduces adrenal weight, serum ACTH and adrenal 5-ene-3 beta-HSD activity. In vitro experiments show that alpha 2u-globulin fails to change adrenal 5-ene-3 beta-HSD activity in corticosterone pretreated rats while in normal and estrogen pretreated rats alpha 2u-globulin increases 5-ene-3 beta-HSD activity.