Merck
  • Acridinic acid: a new antagonist of amino acid-induced excitations of central neurones.
Acridinic acid: a new antagonist of amino acid-induced excitations of central neurones.
Neuroscience letters (1986-05-06)
K Curry, D S Magnuson, H McLennan, M J Peet
ABSTRACT
The actions of acridinic acid (2,3-quinoline dicarboxylic acid), a new derivative of quinolinic acid, as an antagonist of amino acid-induced excitations are described. Acridinate, like kynurenate, in the cerebral cortex reduced the effects of all amino acids equally, but in the spinal cord was significantly less active against quisqualate.
Sigma-Aldrich
3-Hydroxypyridine-2-carboxylic acid, 98%
Supelco
3-Hydroxypicolinic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)