Merck
  • Lumpectomy plus tamoxifen with or without irradiation in women age 70 years or older with early breast cancer: long-term follow-up of CALGB 9343.
Lumpectomy plus tamoxifen with or without irradiation in women age 70 years or older with early breast cancer: long-term follow-up of CALGB 9343.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-05-22)
Kevin S Hughes, Lauren A Schnaper, Jennifer R Bellon, Constance T Cirrincione, Donald A Berry, Beryl McCormick, Hyman B Muss, Barbara L Smith, Clifford A Hudis, Eric P Winer, William C Wood
ABSTRACT
To determine whether there is a benefit to adjuvant radiation therapy after breast-conserving surgery and tamoxifen in women age ≥ 70 years with early-stage breast cancer. Between July 1994 and February 1999, 636 women (age ≥ 70 years) who had clinical stage I (T1N0M0 according to TNM classification) estrogen receptor (ER) -positive breast carcinoma treated by lumpectomy were randomly assigned to receive tamoxifen plus radiation therapy (TamRT; 317 women) or tamoxifen alone (Tam; 319 women). Primary end points were time to local or regional recurrence, frequency of mastectomy, breast cancer-specific survival, time to distant metastasis, and overall survival (OS). Median follow-up for treated patients is now 12.6 years. At 10 years, 98% of patients receiving TamRT (95% CI, 96% to 99%) compared with 90% of those receiving Tam (95% CI, 85% to 93%) were free from local and regional recurrences. There were no significant differences in time to mastectomy, time to distant metastasis, breast cancer-specific survival, or OS between the two groups. Ten-year OS was 67% (95% CI, 62% to 72%) and 66% (95% CI, 61% to 71%) in the TamRT and Tam groups, respectively. With long-term follow-up, the previously observed small improvement in locoregional recurrence with the addition of radiation therapy remains. However, this does not translate into an advantage in OS, distant disease-free survival, or breast preservation. Depending on the value placed on local recurrence, Tam remains a reasonable option for women age ≥ 70 years with ER-positive early-stage breast cancer.
Product Description
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Tamoxifen citrate, European Pharmacopoeia (EP) Reference Standard