Merck
  • [Syntheses of human little gastrin-I and its leucine-15, norleucine-15 and methoxinine-15 analogs].
[Syntheses of human little gastrin-I and its leucine-15, norleucine-15 and methoxinine-15 analogs].
Hoppe-Seyler's Zeitschrift fur physiologische Chemie (1983-02-01)
L Moroder, W Göhring, R Nyfeler, R Scharf, P Thamm, G Wendlberger
ABSTRACT
Human little gastrin-I is known to exhibit a high tendency to air-oxidation of its methionine-15 residue to the corresponding S-oxide derivative, with concomitant loss of biological activity. Since its leucine-15 analog, even if fully biologically active, differs significantly from the parent hormone in the immunological properties, the norleucine-15 and methoxinine-15 analogues were synthetized. For the required comparative analyses new syntheses of human little gastrin-I and of its leucine-15 analog were additionally elaborated. Upon an optimized condensation of the fragments, followed by the deprotection step, partition chromatography as well as preparative high-performance liquid chromatography led to the desired gastrins in satisfactory yields and high degree of purity as judged by the expected and known side products.
Sigma-Aldrich
2-Morpholinoethyl isocyanide, ≥98.0% (GC)
Sigma-Aldrich
[Leu15]-Gastrin I human, ≥95% (HPLC)