Merck
  • A structural snapshot of CYP2B4 in complex with paroxetine provides insights into ligand binding and clusters of conformational states.
A structural snapshot of CYP2B4 in complex with paroxetine provides insights into ligand binding and clusters of conformational states.
The Journal of pharmacology and experimental therapeutics (2013-05-02)
Manish B Shah, Irina Kufareva, Jaime Pascual, Qinghai Zhang, C David Stout, James R Halpert
ABSTRACT
An X-ray crystal structure of CYP2B4 in complex with the drug paroxetine [(3S,4R)-3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine] was solved at 2.14 Å resolution. The structure revealed a conformation intermediate to that of the recently solved complex with amlodipine and that of the more compact complex with 4-(4-chlorophenyl)imidazole in terms of the placement of the F-G cassette. Moreover, comparison of the new structure with 15 previously solved structures of CYP2B4 revealed some new insights into the determinants of active-site size and shape. The 2B4-paroxetine structure is nearly superimposable on a previously solved closed structure in a ligand-free state. Despite the overall conformational similarity among multiple closed structures, the active-site cavity volume of the paroxetine complex is enlarged. Further analysis of the accessible space and binding pocket near the heme reveals a new subchamber that resulted from the movement of secondary structural elements and rearrangements of active-site side chains. Overall, the results from the comparison of all 16 structures of CYP2B4 demonstrate a cluster of protein conformations that were observed in the presence or absence of various ligands.
Product Description
Paroxetine hydrochloride hemihydrate, European Pharmacopoeia (EP) Reference Standard
Paroxetine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paroxetine hydrochloride hemihydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®