Merck
  • The vibration properties of the (n,0) boron nitride nanotubes from ab initio quantum chemical simulations.
The vibration properties of the (n,0) boron nitride nanotubes from ab initio quantum chemical simulations.
The Journal of chemical physics (2013-02-15)
A Erba, M Ferrabone, J Baima, R Orlando, M Rérat, R Dovesi
ABSTRACT
The vibration spectrum of single-walled zigzag boron nitride (BN) nanotubes is simulated with an ab initio periodic quantum chemical method. The trend towards the hexagonal monolayer (h-BN) in the limit of large tube radius R is explored for a variety of properties related to the vibrational spectrum: vibration frequencies, infrared intensities, oscillator strengths, and vibration contributions to the polarizability tensor. The (n,0) family is investigated in the range from n = 6 (24 atoms in the unit cell and tube radius R = 2.5 Å) to n = 60 (240 atoms in the cell and R = 24.0 Å). Simulations are performed using the CRYSTAL program which fully exploits the rich symmetry of this class of one-dimensional periodic systems: 4n symmetry operators for the general (n,0) tube. Three sets of infrared active phonon bands are found in the spectrum. The first one lies in the 0-600 cm(-1) range and goes regularly to zero when R increases; the connection between these normal modes and the elastic and piezoelectric constants of h-BN is discussed. The second (600-800 cm(-1)) and third (1300-1600 cm(-1)) sets tend regularly, but with quite different speed, to the optical modes of the h-BN layer. The vibrational contribution of these modes to the two components (parallel and perpendicular) of the polarizability tensor is also discussed.
Sigma-Aldrich
Boron nitride, powder, ~1 μm, 98%