Merck
  • Orientation correlation of p-nitroaniline molecules in acetone solution observed by hyper-Rayleigh scattering.
Orientation correlation of p-nitroaniline molecules in acetone solution observed by hyper-Rayleigh scattering.
The Journal of chemical physics (2013-02-15)
David P Shelton
ABSTRACT
Measurements of the polarization dependence of hyper-Rayleigh scattering (HRS) by solutions of p-nitroaniline (PNA) in acetone-d6 were used to study molecular orientation correlations. The HRS observations were analyzed in terms of short range direct dipole-dipole correlations between the PNA molecules, a long range transverse polar mode for the PNA molecules, and a long range ion-induced longitudinal polar mode. The conclusion that long range correlations are present is opposite to that in previous work, due to previous neglect of the ion-induced contribution. Depolarized HRS not explained by local correlations appears above 0.1 M PNA concentration, and analysis of the HRS observations indicates strong orientation correlations with a range of 10 nm for the PNA molecules.
Product Description
Supelco
Acetone, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, natural, ≥97%
Pricing and availability is not currently available.
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Nitroaniline, ≥99%
Pricing and availability is not currently available.
Supelco
4-Nitroaniline, analytical standard
Pricing and availability is not currently available.