Merck
  • Structural studies of the pectic polysaccharide from Siberian fir (Abies sibirica Ledeb.).
Structural studies of the pectic polysaccharide from Siberian fir (Abies sibirica Ledeb.).
Carbohydrate polymers (2013-02-13)
Elena N Makarova, Olga A Patova, Evgeny G Shakhmatov, Sergey P Kuznetsov, Yury S Ovodov
ABSTRACT
The pectic polysaccharide named abienan AS-A was isolated from the wood greenery of Abies sibirica using dilute hydrochloric acid (pH 4.0) at 70°C. The structure of abienan AS-A was elucidated using sugar composition analysis, ion-exchange chromatography and partial acid hydrolysis followed by NMR spectroscopy. The linear region of abienan AS-A was shown to contain linear 1,4-α-D-galactopyranosyluronan partially substituted with methyl esters or 3-O-acetyl groups and rhamnogalacturonan blocks consisting of 1,4-α-D-galacturonan partially substituted with methyl ester groups and connected by 2-O-substituted α-rhamnopyranose residues. The branched region of abienan AS-A was found to be made of RG-I. The side chains of RG-I were shown to contain 1,4-β-galactan and branched arabinan. Some 4-O-substituted β-galactopyranose residues were shown to be attached to the 4-position of the 2-O-substituted α-rhamnopyranose residues of the RG-I backbone. The arabinan groups were made up of a 1,5-linked α-L-arabinofuranan backbone that was 3-O-, 2-O-, and 2,3-di-O-substituted with the terminal and 1,3-linked α-L-arabinofuranose residues.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur