- Efficacy of amitraz-impregnated collars on white-tailed deer (Artiodactyla: Cervidae) in reducing free-living populations of lone star ticks (Acari: Ixodidae).
Efficacy of amitraz-impregnated collars on white-tailed deer (Artiodactyla: Cervidae) in reducing free-living populations of lone star ticks (Acari: Ixodidae).
Over a 7 yr period, we monitored the effect of a commercially available, amitraz impregnated anti-tick collar in controlling free-living populations of lone star ticks, Amblyomma americanum (L.) when manually fitted around necks of white-tailed deer, Odocoileus virginianus (Zimmermann). Study animals in treatment and control groups were confined in 38.8 ha game-fenced and densely vegetated treatment plots in South Texas. Tick densities during years 1 and 7 served as untreated pre- and posttreatment comparisons and treatments occurred during years 2 through 5. Reductions in tick densities in the treatment plot were compared against tick densities in a control plot having similar vegetation and numbers of untreated deer. During years of treatment, indices of control pressure ranged from 18.2 to 82.6 for nymphs and 16.9-78.7 for adults, and efficacy, expressed as percentage control during the final year of treatment, was 77.2 and 85.0%, respectively, for nymphal and adult ticks. These data show that acaricidal collar treatments provide efficacies very similar to those achieved with the existing ivermectin-medicated bait and '4-Poster' topical treatment technologies to control ticks feeding on wild white-tailed deer.