Merck
  • Polyamino geranic derivatives as new chemosensitizers to combat antibiotic resistant gram-negative bacteria.
Polyamino geranic derivatives as new chemosensitizers to combat antibiotic resistant gram-negative bacteria.
Bioorganic & medicinal chemistry (2013-01-29)
Jean Michel Brunel, Aurélie Lieutaud, Vincent Lome, Jean-Marie Pagès, Jean-Michel Bolla
ABSTRACT
Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate reagent (BOP) serves as an efficient and versatile coupling reagent for the design and synthesis of new polyamino geranic acid derivatives in moderate to good chemical yields varying from 47% to 83%. These compounds induced a significant decrease of antibiotic resistance in two Gram-negative bacterial MDR strains. Our data suggested that their mechanism of action is closely associated with the inhibition of the efflux pumps.
Sigma-Aldrich
Geranic acid, 85%, stabilised
Pricing and availability is not currently available.
Sigma-Aldrich
Geranic acid, technical grade, 85%
Pricing and availability is not currently available.