Merck
  • Protective role of quercetin on polychlorinated biphenyls (Aroclor-1254) induced oxidative stress and apoptosis in liver of adult male rats.
Protective role of quercetin on polychlorinated biphenyls (Aroclor-1254) induced oxidative stress and apoptosis in liver of adult male rats.
Journal of biochemical and molecular toxicology (2013-01-03)
Suganya Sekaran, Selvakumar Kandaswamy, Krishnamoorthy Gunasekaran, Elumalai Perumal, Fariya Yasmine Afsar Basha, Balasakthi Janani Madhan Mohan, Arunakaran Jagadeesan
ABSTRACT
The present study aims to investigate the protective effect of quercetin against Aroclor-1254-induced hepatotoxicity in rats. Male Wistar rats were grouped into Group I control received vehicle (corn oil; 1 mL/kg bwt); Group II quercetin alone (50 mg/kg bwt/day orally); Group III Aroclor-1254 (2 mg/kg bwt/day intraperitoneally); Group IV Aroclor-1254 + quercetin treated for 30 days. The Aroclor-1254 treatment caused significant alteration in the biochemical parameters (hydrogen peroxide, lipid peroxidation, reduced glutathione levels, and alkaline phosphatase activity). The expressions of apoptotic and antiapoptotic proteins and the liver histology of Aroclor-1254-exposed rats showed cytoplasmic degeneration along with infiltration of polymorphonuclear cells. Whereas simultaneous treatment with quercetin normalized all the biochemical parameters, consequently it inhibited apoptosis mediated by Aroclor-1254 by downregulating aryl hydrocarbon receptor, p53 and apoptotic protein (Bax, caspase-9, caspase-3) and upregulating the antiapoptotic protein (Bcl-2) expression patterns; thereby, quercetin reduces alteration in hepatocellular morphology. Thus quercetin exhibited hepatoprotective effect.
Product Description
Supelco
Aroclor 1254 solution, certified reference material, 500 mg/kg in transformer oil
Supelco
Aroclor 1254 solution, certified reference material, 50 mg/kg in transformer oil
Supelco
Aroclor 1254, ampule of 50 mg, analytical standard