Merck
  • Profiling the evanescent field of nanofiber waveguides using self-assembled polymer coatings.
Profiling the evanescent field of nanofiber waveguides using self-assembled polymer coatings.
Nanoscale (2012-12-13)
Ilsun Yoon, Sarah E Baker, Kanguk Kim, Yinmin Wang, Sadik C Esener, Donald J Sirbuly
ABSTRACT
Here we demonstrate a facile method of quantifying the decaying optical field surrounding free-standing tin dioxide (SnO(2)) nanofiber waveguides. Through the use of thin self-assembled polyelectrolyte coatings and fluorescent optical transmitters we map out the optical intensity as a function of distance with nanometer resolution.
Sigma-Aldrich
Tin(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size
Sigma-Aldrich
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis