Merck
  • Development of poly(vinyl acetate-methylacrylic acid)/chitosan/Fe3O4 nanoparticles for the diagnosis of non-alcoholic steatohepatitis with magnetic resonance imaging.
Development of poly(vinyl acetate-methylacrylic acid)/chitosan/Fe3O4 nanoparticles for the diagnosis of non-alcoholic steatohepatitis with magnetic resonance imaging.
Journal of materials science. Materials in medicine (2012-09-15)
Xiadan Luo, Xiaoli Song, Aiping Zhu, Yunfeng Si, Lijun Ji, Zhanrong Ma, Zhiyun Jiao, Jingtao Wu
ABSTRACT
Non-alcoholic steatohepatitis is a burgeoning health problem. To diagnose NASH with magnetic resonance imaging (MRI), an effective contrast agent, a stable suspension of superparamagnetic Fe(3)O(4) nanoparticles, were newly developed. The negatively charged Fe(3)O(4) nanoparticles were coated with positive chitosan (CS) firstly, and then assembled with poly(vinyl acetate-methylacrylic acid) (P(VAc-MAA)). Transmission electron microscope and dynamic light scattering confirmed that the obtained P(VAc-MAA)/CS/Fe(3)O(4) nanoparticles had a spherical or ellipsoidal morphology with an average diameter in the range of 14-20 nm. The superparamagnetic property and spinel structure of the Fe(3)O(4) nanoparticles were well preserved due to the protection of the P(VAc-MAA)/CS layers on the surface of the Fe(3)O(4) nanoparticles. The in vivo rat experiments confirmed that the P(VAc-MAA)/CS/Fe(3)O(4) nanoparticles were an effective contrast agent for MRI to diagnose NASH.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Zinc acrylate, 98%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Vinyl acetate solution, certified reference material, 5000 μg/mL in acetonitrile
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Vinyl acetate, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Vinyl acetate, analytical standard