- On the aromatic stabilization of benzenoid hydrocarbons.
On the aromatic stabilization of benzenoid hydrocarbons.
Chemical communications (Cambridge, England) (2012-09-11)
Arkadiusz Ciesielski, Dorota K Stepień, Michał A Dobrowolski, Łukasz Dobrzycki, Michał K Cyrański
PMID22962659
ABSTRACT
A general scheme for estimation of aromatic stabilization energies of benzenoid hydrocarbons based on selected topological features has been presented. The reactions have been applied to benzene, naphthalene, anthracene, phenanthrene, pyrene, tetracene, benz[a]anthracene, chrysene, [4]-helicene, anthanthrene and coronene.
Product Description
Coronene, BCR®, certified reference material
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Chrysene, BCR®, certified reference material
Anthanthrene, BCR®, certified reference material