Merck
  • Cerebrospinal fluid-based kinetic biomarkers of axonal transport in monitoring neurodegeneration.
Cerebrospinal fluid-based kinetic biomarkers of axonal transport in monitoring neurodegeneration.
The Journal of clinical investigation (2012-08-28)
Patrizia Fanara, Po-Yin A Wong, Kristofor H Husted, Shanshan Liu, Victoria M Liu, Lori A Kohlstaedt, Timothy Riiff, Joan C Protasio, Drina Boban, Salena Killion, Maudi Killian, Lorrie Epling, Elisabeth Sinclair, Julia Peterson, Richard W Price, Deborah E Cabin, Robert L Nussbaum, Jörg Brühmann, Roland Brandt, Chadwick W Christine, Michael J Aminoff, Marc K Hellerstein
ABSTRACT
Progress in neurodegenerative disease research is hampered by the lack of biomarkers of neuronal dysfunction. We here identified a class of cerebrospinal fluid-based (CSF-based) kinetic biomarkers that reflect altered neuronal transport of protein cargo, a common feature of neurodegeneration. After a pulse administration of heavy water (2H2O), distinct, newly synthesized 2H-labeled neuronal proteins were transported to nerve terminals and secreted, and then appeared in CSF. In 3 mouse models of neurodegeneration, distinct 2H-cargo proteins displayed delayed appearance and disappearance kinetics in the CSF, suggestive of aberrant transport kinetics. Microtubule-modulating pharmacotherapy normalized CSF-based kinetics of affected 2H-cargo proteins and ameliorated neurodegenerative symptoms in mice. After 2H2O labeling, similar neuronal transport deficits were observed in CSF of patients with Parkinson's disease (PD) compared with non-PD control subjects, which indicates that these biomarkers are translatable and relevant to human disease. Measurement of transport kinetics may provide a sensitive method to monitor progression of neurodegeneration and treatment effects.
Sigma-Aldrich
(S,R)-Noscapine, 97%