Merck
  • New glycosides of acetophenone derivatives and phenylpropanoids from Juniperus occidentalis.
New glycosides of acetophenone derivatives and phenylpropanoids from Juniperus occidentalis.
Journal of natural medicines (2012-08-01)
Yuka Inatomi, Hiroko Murata, Akira Inada, Tsutomu Nakanishi, Frank A Lang, Jin Murata, Munekazu Iinuma
ABSTRACT
New glycosides of seven acetophenone derivatives (1-7) and two phenylpropanoids (8, 9), named juniperosides III-XI, have been isolated from the MeOH extract of the leaves and stems of Juniperus occidentalis Hook. (Cupressaceae), together with eleven other known compounds. The structures of these compounds have been successfully elucidated using a variety of spectroscopic techniques.
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Acetophenone, ≥98%, FG
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%