Merck
  • Hsp90 stabilizes Cdc25A and counteracts heat shock-mediated Cdc25A degradation and cell-cycle attenuation in pancreatic carcinoma cells.
Hsp90 stabilizes Cdc25A and counteracts heat shock-mediated Cdc25A degradation and cell-cycle attenuation in pancreatic carcinoma cells.
Human molecular genetics (2012-07-31)
Benedikt Giessrigl, Sigurd Krieger, Margit Rosner, Nicole Huttary, Philipp Saiko, Mouad Alami, Samir Messaoudi, Jean-François Peyrat, Alexandre Maciuk, Michaela Gollinger, Sabine Kopf, Egidijus Kazlauskas, Peter Mazal, Thomas Szekeres, Markus Hengstschläger, Daumantas Matulis, Walter Jäger, Georg Krupitza
ABSTRACT
Pancreas cancer cells escape most treatment options. Heat shock protein (Hsp)90 is frequently over-expressed in pancreas carcinomas and protects a number of cell-cycle regulators such as the proto-oncogene Cdc25A. We show that inhibition of Hsp90 with geldanamycin (GD) destabilizes Cdc25A independent of Chk1/2, whereas the standard drug for pancreas carcinoma treatment, gemcitabine (GEM), causes Cdc25A degradation through the activation of Chk2. Both agents applied together additively inhibit the expression of Cdc25A and the proliferation of pancreas carcinoma cells thereby demonstrating that both Cdc25A-destabilizing/degrading pathways are separated. The role of Hsp90 as stabilizer of Cdc25A in pancreas carcinoma cells is further supported by two novel synthetic inhibitors 4-tosylcyclonovobiocic acid and 7-tosylcyclonovobiocic acid and specific Hsp90AB1 (Hsp90β) shRNA. Our data show that targeting Hsp90 reduced the resistance of pancreas carcinoma cells to treatment with GEM.
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Supelco
Novobiocin sodium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Novobiocin sodium, meets USP testing specifications
Sigma-Aldrich
Novobiocin sodium salt, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid