Merck
  • [Immobilization of a recombinant strain producing glucose isomerase on SiO2-xerogel and properties of prepared biocatalysts].
[Immobilization of a recombinant strain producing glucose isomerase on SiO2-xerogel and properties of prepared biocatalysts].
Prikladnaia biokhimiia i mikrobiologiia (2011-03-01)
G A Kovalenko, L V Perminova, T V Chuenko, L I Sapunova, E A Shliakhotko, A G Lobanok
ABSTRACT
An original method of immobilization of nongrowing microorganism cells on xerogel of silicon dioxide containing insoluble hydroxyl compounds of cobalt(III) has been developed. A recombinant strain producing glucose isomerase has been constructed on the basis of Escherichia coli with the use of a gene of Arthrobacter nicotianae. It was revealed that glucose isomerase activity and stability of biocatalysts prepared on the basis of the recombinant E. coli strain was 3-5 times greater compared with the biocatalysts prepared with the use of the donor strain A. nicotianae. Under conditions of continuous hydrolysis of 3 M fructose at 62-65 degrees C in a fixed bed reactor, time of half-inactivation of the biocatalysts prepared from the recombinant strain and A. nicotianae was -60 and -25 days, respectively.
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, reagent grade, 98%
Sigma-Aldrich
Cobalt(II) nitrate hexahydrate, ACS reagent, ≥98%