Merck
  • In situ catalytic pyrolysis of lignocellulose using alkali-modified amorphous silica alumina.
In situ catalytic pyrolysis of lignocellulose using alkali-modified amorphous silica alumina.
Bioresource technology (2012-06-19)
M Zabeti, T S Nguyen, L Lefferts, H J Heeres, K Seshan
ABSTRACT
Canadian pinewood was pyrolyzed at 450 °C in an Infrared oven and the pyrolysis vapors were converted by passing through a catalyst bed at 450 °C. The catalysts studied were amorphous silica alumina (ASA) containing alkali metal or alkaline earth metal species including Na, K, Cs, Mg and Ca. The catalysts effectiveness to reduce the bio-oil oxygen content, to enhance the bio-oil energy density and to change the liquid and gas product distribution were evaluated using different techniques including gravimetric analysis, elemental analysis, Karl-Fischer titration, GC/MS and micro-GC analysis. According to the results K/ASA found to be the most effective catalysts for conversion of hollocellulose (hemicellulose and cellulose)-derived vapors of pinewood while Cs/ASA catalyst was the most effective catalyst for conversion of lignin-derived vapors and production of hydrocarbons.
Sigma-Aldrich
Drying pearls orange