Merck
  • Topical silver sulfadiazine vs collagenase ointment for the treatment of partial thickness burns in children: a prospective randomized trial.
Topical silver sulfadiazine vs collagenase ointment for the treatment of partial thickness burns in children: a prospective randomized trial.
Journal of pediatric surgery (2012-06-19)
Daniel J Ostlie, David Juang, Pablo Aguayo, Janine P Pettiford-Cunningham, Erin A Erkmann, Diane E Rash, Susan W Sharp, Ronald J Sharp, Shawn D St Peter
ABSTRACT
The 2 most commonly used topical agents for partial thickness burns are silver sulfadiazine (SSD) and collagenase ointment (CO). Silver sulfadiazine holds antibacterial properties, and eschar separation occurs naturally. Collagenase ointment is an enzyme that cleaves denatured collagen facilitating separation but has no antibacterial properties. Currently, there are no prospective comparative data in children for these 2 agents. Therefore, we conducted a prospective randomized trial. After institutional review board approval, patients were randomized to daily debridement with SSD or CO. Primary outcome was the need for skin grafting. Patients were treated for 2 days with SSD with subsequent randomization. Polymyxin was mixed with CO for antibacterial coverage. Debridements were performed daily for 10 days or until the burn healed. Grafting was performed after 10 days if not healed. From January 2008 to January 2011, 100 patients were enrolled, with no differences in patient characteristics. There were no differences in clinical course, outcome, or need for skin grafting. Wound infections occurred in 7 patients treated with CO and 1 patient treated with SSD (P = .06). Collagenase ointment was more expensive than SSD (P < .001). However, total hospital charges did not differ. There are no differences in outcomes between topical SSD or CO in the management of childhood burns results.
Sigma-Aldrich
Silver(I) sulfadiazine, 98%