Merck
  • Biosynthesis of α- and β-ionone, prominent scent compounds, in flowers of Osmanthus fragrans.
Biosynthesis of α- and β-ionone, prominent scent compounds, in flowers of Osmanthus fragrans.
Acta biochimica Polonica (2012-03-20)
Susanne Baldermann, Masaya Kato, Peter Fleischmann, Naoharu Watanabe
ABSTRACT
Carotenoid derived volatiles are important fragrance compounds, which contribute to the scents of flowers from diverse taxa. A famous example is represented by the flowers of Osmanthus fragrans where apocarotenoids account for more than 20% of all volatiles. In the recent years, bio-degradation of carotenoids has been shown to be an important route for apocarotenoids formation. Here, we report on the contribution the O. fragrans carotenoid cleavage dioxygenase 1 to the synthesis of the two predominant C(13)-apocarotenoids, α- and β-ionone, derived from α-and β-carotene, respectively.
Sigma-Aldrich
β-Ionone, natural, ≥95%, FG
Sigma-Aldrich
β-Ionone, predominantly trans, ≥97%, FCC, FG
Sigma-Aldrich
β-Ionone, 96%
Sigma-Aldrich
α-Ionone, ≥90%, stabilized
Sigma-Aldrich
α-Ionone, technical grade, 90%