Merck
  • CCK-B receptor gene and response to cholecystokinin-tetrapeptide in healthy volunteers.
CCK-B receptor gene and response to cholecystokinin-tetrapeptide in healthy volunteers.
Peptides (2012-03-15)
Diana Koszycki, Zoe Prichard, Alexandra J Fiocco, Jakov Shlik, James L Kennedy, Jacques Bradwejn
ABSTRACT
Recent investigations suggest that genes that confer risk for panic disorder (PD) may moderate response to panicogenic agents in healthy volunteers. Given the potential role of the central cholecystokinin receptor (CCKBR) (CT) polymorphism alleles 26 and 27 in PD, the present study attempted to discern if these alleles moderated panicogenic sensitivity to the CCKBR agonist, CCK-tetrapeptide (CCK-4), in healthy volunteers. The study group consisted of 92 men and women with no personal or family history of psychiatric illness. Participants provided blood samples for genotyping of the CCKBR alleles and they received a 25 μg bolus injection of CCK-4. Behavioral, cardiovascular and hormonal responses to the peptide were assessed and analyzed with adjusted linear regression models. Carriers of the CCKBR alleles tended to have higher levels of pre-challenge anxiety and significantly higher levels of anxiety sensitivity and introversion than those without the alleles. However, they did not exhibit an enhanced panicogenic response to CCK-4. Overall, our findings do not demonstrate a role of these alleles in modulating CCK-4's panicogenicity. The significant association between the risk alleles and anxiety-related personality traits is intriguing and further exploration of this association is merited.
Sigma-Aldrich
Cholecystokinin Fragment 30-33 Amide, ≥95% (HPLC)