Merck
  • Elevation of GM2 ganglioside during ethanol-induced apoptotic neurodegeneration in the developing mouse brain.
Elevation of GM2 ganglioside during ethanol-induced apoptotic neurodegeneration in the developing mouse brain.
Journal of neurochemistry (2012-03-01)
Mitsuo Saito, Goutam Chakraborty, Relish Shah, Rui-Fen Mao, Asok Kumar, Dun-Sheng Yang, Kostantin Dobrenis, Mariko Saito
ABSTRACT
GM2 ganglioside in the brain increased during ethanol-induced acute apoptotic neurodegeneration in 7-day-old mice. A small but a significant increase observed 2 h after ethanol exposure was followed by a marked increase around 24 h. Subcellular fractionation of the brain 24 h after ethanol treatment indicated that GM2 increased in synaptic and non-synaptic mitochondrial fractions as well as in a lysosome-enriched fraction characteristic to the ethanol-exposed brain. Immunohistochemical staining of GM2 in the ethanol-treated brain showed strong punctate staining mainly in activated microglia, in which it partially overlapped with staining for LAMP1, a late endosomal/lysosomal marker. Also, there was weaker neuronal staining, which partially co-localized with complex IV, a mitochondrial marker, and was augmented in cleaved caspase 3-positive neurons. In contrast, the control brain showed only faint and diffuse GM2 staining in neurons. Incubation of isolated brain mitochondria with GM2 in vitro induced cytochrome c release in a manner similar to that of GD3 ganglioside. Because ethanol is known to trigger mitochondria-mediated apoptosis with cytochrome c release and caspase 3 activation in the 7-day-old mouse brain, the GM2 elevation in mitochondria may be relevant to neuroapoptosis. Subsequently, activated microglia accumulated GM2, indicating a close relationship between GM2 and ethanol-induced neurodegeneration.
Sigma-Aldrich
Monosialoganglioside GM2 from bovine brain, ≥95% (TLC)