Merck
  • Simultaneous purification and site-specific modification of pyrroline-carboxy-lysine proteins.
Simultaneous purification and site-specific modification of pyrroline-carboxy-lysine proteins.
Chembiochem : a European journal of chemical biology (2012-01-10)
Hsien-Po Chiu, Jan Grünewald, Xueshi Hao, Ansgar Brock, Linda Okach, Tetsuo Uno, Bernhard H Geierstanger
ABSTRACT
Sticky residue: Pyrroline-carboxy-lysine (Pcl) can be readily incorporated into proteins expressed in E. coli and mammalian cells by using the pyrrolysyl tRNA/tRNA synthetase pair. Pcl can be used as a single amino acid purification tag and can be site-specifically modified with functional probes during the elution process.
Sigma-Aldrich
2-Aminobenzaldehyde, ≥98%