Merck
  • Main odorants in Jura flor-sherry wines. Relative contributions of sotolon, abhexon, and theaspirane-derived compounds.
Main odorants in Jura flor-sherry wines. Relative contributions of sotolon, abhexon, and theaspirane-derived compounds.
Journal of agricultural and food chemistry (2011-11-29)
Sonia Collin, Sabrina Nizet, Thomas Claeys Bouuaert, Pierre-Marie Despatures
ABSTRACT
The aromatic profile of Jura flor-sherry wines (also called "yellow wines") has been little studied. Only acetaldehyde, diethoxy-1,1-ethane, and sotolon have been described as key odorants. In the present work, three wines (vintages 2002 and 2003) were investigated by gas chromatography-mass spectrometry and gas chromatography-olfactometry (GC-O) aroma extract dilution analysis. The goal was to assess the relative impact of varietal, fermentation, and oak-barrel compounds by using two complementary extraction procedures. No grape terpenoids were found after the long barrel aging (6 years and 3 months). On the other hand, two candy/fruity esters issued from yeast exhibited high flavor dilution factor (FD) values: ethyl isobutyrate (64-1024) and ethyl isovalerate (128-1024). As expected, many oak-related odorants were found in the XAD 2 flavor extracts, mainly homofuraneol [2-ethyl-4-hydroxy-5-methyl-3(2H)-furanone] (cotton candy, FD = 16-256) and cis-β-methyloctalactone (butter, woody, FD = 256). Most probably issued from oxidation of the grape constituent theaspirane, an exceptional grenadine odor was perceived by GC-O up to dilution 64-1024. Chemical oxidation experiments and GC-high-resolution mass spectrometry (HRMS) allowed us to identify it as 4-hydroxy-7,8-dihydro-β-ionone (RI(CPsil5CB) = 1373), a hydrolysis-derived product of dihydrodehydro-β-ionone. With an extraction dedicated to hydrophilic compounds, the key role of sotolon was confirmed (112-387 μg/kg; FD = 256-1024). This procedure enabled us to also evidence its ethyl analogue, abhexon (31-74 μg/kg; FD = 64-256).
Sigma-Aldrich
4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, ≥97%, FG
Supelco
4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, analytical standard
Sigma-Aldrich
Ethyl isobutyrate, 99%