MilliporeSigma
  • Flunarizine inhibits sensory neuron excitability by blocking voltage-gated Na+ and Ca2+ currents in trigeminal ganglion neurons.
Flunarizine inhibits sensory neuron excitability by blocking voltage-gated Na+ and Ca2+ currents in trigeminal ganglion neurons.
Chinese medical journal (2011-11-02)
Qing Ye, Qiang Wang, Lan-yun Yan, Wen-hui Wu, Sha Liu, Hang Xiao, Qi Wan
ABSTRACT
Although flunarizine has been widely used for migraine prophylaxis with clear success, the mechanisms of its actions in migraine prophylaxis are not completely understood. The aim of this study was to investigate the effects of flunarizine on tetrodotoxin-resistant Na(+) channels and high-voltage activated Ca(2+) channels of acutely isolated mouse trigeminal ganglion neurons. Sodium currents and calcium currents in trigeminal ganglion neurons were monitored using whole-cell patch-clamp recordings. Paired Student's t test was used as appropriate to evaluate the statistical significance of differences between two group means. Both tetrodotoxin-resistant sodium currents and high-voltage activated calcium currents were blocked by flunarizine in a concentration-dependent manner with the concentration producing half-maximal current block values of 2.89 µmol/L and 2.73 µmol/L, respectively. The steady-state inactivation curves of tetrodotoxin-resistant sodium currents and high-voltage activated calcium currents were shifted towards more hyperpolarizing potentials after exposure to flunarizine. Furthermore, the actions of flunarizine in blocking tetrodotoxin-resistant sodium currents and high-voltage activated calcium currents were use-dependent, with effects enhanced at higher rates of channel activation. Blockades of these currents might help explain the peripheral mechanism underlying the preventive effect of flunarizine on migraine attacks.
Sigma-Aldrich
Flunarizine dihydrochloride, ≥98% (TLC)