Merck
  • Removal of polycyclic aromatic hydrocarbons from aqueous solution on soybean stalk-based carbon.
Removal of polycyclic aromatic hydrocarbons from aqueous solution on soybean stalk-based carbon.
Journal of environmental quality (2011-10-28)
Huoliang Kong, Jiao He, Yanzheng Gao, Jin Han, Xuezhu Zhu
ABSTRACT
Soybean [ (L.) Merr.] stalk-based carbons were prepared by phosphoric acid activation at different carbonization temperatures. Characteristics of the prepared carbon, including specific surface area, iodine number, and amount of methylene blue sorption, were determined. Experiments on phenanthrene, naphthalene, and acenaphthene, as representatives of polycyclic aromatic hydrocarbons (PAHs), removal from aqueous solution by the prepared carbon were conducted at different levels of carbon addition. The results indicated that the specific surface area, iodine number, and amount of methylene blue sorption increased with an increase of carbonization temperature. The maximum values were observed at 700°C and were 287.63 m g, 508.99 mg g, and 90.14 mg g, respectively. The removal efficiencies of phenanthrene, naphthalene, and acenaphthene tended to increase with increasing carbon amounts and carbonization temperature. The optimal removal performance was obtained under the experimental conditions of carbon concentrations of 0.04 g 32 mL and carbonization temperature of 700°C, and the removal efficiencies of phenanthrene, naphthalene, and acenaphthene were 99.89, 100, and 95.64%, respectively. The performance of the prepared carbon was superior to that of commercial activated carbon. Additionally, for the same carbon concentrations, the removal efficiency of PAHs on prepared carbons followed the order: phenanthrene > naphthalene > acenaphthene. Results obtained from this work provide some insight into the reuse of an agricultural residue, and also provide a new application for the treatment of PAHs in contaminated water utilizing activated carbon prepared from agricultural residues.
Product Description
Supelco
Acenaphthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acenaphthene, analytical standard
Sigma-Aldrich
Acenaphthene, 99%