Merck
  • Identification of epoxide functionalities in protonated monofunctional analytes by using ion/molecule reactions and collision-activated dissociation in different ion trap tandem mass spectrometers.
Identification of epoxide functionalities in protonated monofunctional analytes by using ion/molecule reactions and collision-activated dissociation in different ion trap tandem mass spectrometers.
Journal of the American Society for Mass Spectrometry (2011-10-18)
Ryan J Eismin, Mingkun Fu, Sonoeun Yem, Fanny Widjaja, Hilkka I Kenttämaa
ABSTRACT
A mass spectrometric method has been delineated for the identification of the epoxide functionalities in unknown monofunctional analytes. This method utilizes gas-phase ion/molecule reactions of protonated analytes with neutral trimethyl borate (TMB) followed by collision-activated dissociation (CAD) in an ion trapping mass spectrometer (tested for a Fourier-transform ion cyclotron resonance and a linear quadrupole ion trap). The ion/molecule reaction involves proton transfer from the protonated analyte to TMB, followed by addition of the analyte to TMB and elimination of methanol. Based on literature, this reaction allows the general identification of oxygen-containing analytes. Vinyl and phenyl epoxides can be differentiated from other oxygen-containing analytes, including other epoxides, based on the loss of a second methanol molecule upon CAD of the addition/methanol elimination product. The only other analytes found to undergo this elimination are some amides but they also lose O = B-R (R = group bound to carbonyl), which allows their identification. On the other hand, other epoxides can be differentiated from vinyl and phenyl epoxides and from other monofunctional analytes based on the loss of (CH(3)O)(2)BOH or formation of protonated (CH(3)O)(2)BOH upon CAD of the addition/methanol elimination product. For propylene oxide and 2,3-dimethyloxirane, the (CH(3)O)(2)BOH fragment is more basic than the hydrocarbon fragment, and the diagnostic ion (CH(3)O)(2)BOH (2) (+) is formed. These reactions involve opening of the epoxide ring. The only other analytes found to undergo (CH(3)O)(2)BOH elimination are carboxylic acids, but they can be differentiated from the rest based on several published ion/molecule reaction methods. Similar results were obtained in the Fourier-transform ion cyclotron resonance and linear quadrupole ion trap mass spectrometer.
Sigma-Aldrich
Ethyl vinyl ether, contains 0.1% KOH as stabilizer, 99%
Sigma-Aldrich
Trimethyl borate, purified by redistillation, ≥99.5%
Sigma-Aldrich
Trimethyl borate, ≥98%
Sigma-Aldrich
Trimethyl borate, purum, ≥99.0% (GC)
Sigma-Aldrich
Trimethyl borate, 99.999% (trace metal basis)