Merck
  • Tailored 3D interface for efficiency improvement in encapsulation-free hybrid light-emitting diodes.
Tailored 3D interface for efficiency improvement in encapsulation-free hybrid light-emitting diodes.
ACS applied materials & interfaces (2011-08-03)
Eugenia Martínez-Ferrero, Amparo Forneli, Cédric Boissière, David Grosso, Clément Sanchez, Emilio Palomares
ABSTRACT
The modification of the planar organic/inorganic interface of the hybrid light-emitting diodes by the addition of a tailored nanoporous crystalline metal oxide increases up to 1 order of magnitude the efficiency of the resulting devices. In this Letter, we present the preparation and characterization of the novel controlled 3D interface and discuss how the interaction between the metal oxide and the polymer at the interface results in such an improvement.
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%