Merck
  • Optimisation of a semiochemical slow-release alginate formulation attractive towards Aphidius ervi Haliday parasitoids.
Optimisation of a semiochemical slow-release alginate formulation attractive towards Aphidius ervi Haliday parasitoids.
Pest management science (2011-07-05)
Stéphanie Heuskin, Stéphanie Lorge, Bruno Godin, Pascal Leroy, Isabelle Frère, François J Verheggen, Eric Haubruge, Jean-Paul Wathelet, Michèle Mestdagh, Thierry Hance, Georges Lognay
ABSTRACT
Optimisation of alginate formulations is described in order to develop semiochemical (E-β-farnesene and E-β-caryophyllene) slow-release devices in biological control approaches by attracting predators and parasitoids of aphids. Various formulation criteria were optimised with respect to semiochemical encapsulation capacity. Moreover, the optimised formulation was characterised by texturometry and confocal microscopy. The slow-release rates of semiochemicals were calculated in laboratory controlled conditions. The attractiveness of semiochemical formulations towards Aphidius ervi was demonstrated by olfactometry. Two major parameters were highlighted in encapsulation optimisation: the type of alginate (Sigma L) and the type of crosslinker ion (Ca(2+)). Other formulation parameters were optimised: ionic strength (0.5 M), Ca(2+) (0.2 M) and alginate (1.5%) concentrations and the maturation time of beads in CaCl(2) solution (48 h). After physical characterisation of beads, semiochemical slow-release measurements showed that alginate formulations were efficient sesquiterpene releasers, with 503 µg of E-β-farnesene and 1791 µg of E-β-caryophyllene totally released in 35 days. The efficiency of semiochemical alginate beads as attractants for female parasitoids was demonstrated, with high percentages of attraction for semiochemical odours (88 and 90% for E-β-farnesene and E-β-caryophyllene respectively) and significant statistical results. Semiochemical alginate beads can be considered as efficient slow-release systems in biological control. These formulations could be very useful to attract aphid parasitoids on crop fields.
Product Description
Supelco
trans-β-Farnesene, analytical standard