Merck
Effect of environmental contaminants on Beta cell function.
International journal of toxicology (2011-06-28)
Emilija Makaji, Sandeep Raha, Michael G Wade, Alison C Holloway
ABSTRACT
There is an increasing concern that chemicals in the environment are contributing to the global rise in the prevalence of type 2 diabetes (T2D). However, there is limited evidence for direct effects of these chemicals on beta cell function. Therefore, the goals of this study were (1) to test the hypothesis that environmental contaminants can directly affect beta cell function and (2) examine mechanistic pathways by which these contaminants could affect beta cell function. Using mouse beta TC-6 cells, we examined the acute effects of 6 substances (benzo[a]pyrene, bisphenol A [BPA], propylparaben, methylparaben, perfluorooctanoic acid, and perfluorooctyl sulfone) on insulin secretion. Only BPA treatment directly affected insulin secretion. Furthermore, chronic exposure to BPA altered the expression of key proteins in the cellular and endoplasmic reticulum stress response. These data suggest that long-term BPA exposure may be detrimental to beta cell function and ultimately be an important contributor to the etiology of T2D.
Product Description
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Methyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
Methyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.