Merck
  • Enhanced topical and transdermal delivery of antineoplastic and antiviral acyclic nucleoside phosphonate cPr-PMEDAP.
Enhanced topical and transdermal delivery of antineoplastic and antiviral acyclic nucleoside phosphonate cPr-PMEDAP.
Pharmaceutical research (2011-06-15)
Kateřina Vávrová, Petra Kovaříková, Barbora Skolová, Martina Líbalová, Jaroslav Roh, Robert Cáp, Antonín Holý, Alexandr Hrabálek
ABSTRACT
Acyclic nucleoside phosphonates possess unique antiviral and antineoplastic activities; however, their polar phosphonate moiety is associated with low ability to cross biological membranes. We explored the potential of transdermal and topical delivery of 2,6-diaminopurine derivative cPr-PMEDAP. In vitro diffusion of cPr-PMEDAP was investigated using formulations at different pH and concentration and with permeation enhancer through porcine and human skin. Ability of 0.1-5% cPr-PMEDAP to cross human skin barrier was very low with flux values ~40 ng/cm(2)/h, the majority of compound found in the stratum corneum. The highest permeation rates were found at pH 6; increased donor concentration had no influence. The permeation enhancer dodecyl 6-dimethylaminohexanoate (DDAK, 1%) increased flux of cPr-PMEDAP (up to 61 times) and its concentration in nucleated epidermis (up to ~0.5 mg of cPr-PMEDAP/g of the tissue). No deamination of cPr-PMEDAP into PMEG occurred during permeation studies, but N-dealkylation into PMEDAP mediated by skin microflora was observed. Transdermal or topical application of cPr-PMEDAP enabled by the permeation enhancer DDAK may provide an attractive alternative route of administration of this potent antitumor and antiviral compound.
Product Description
Supelco
1-Dodecanol, Selectophore, ≥98.0%
Sigma-Aldrich
Lauryl alcohol, ≥98%, FG
Sigma-Aldrich
1-Dodecanol, reagent grade, 98%
Sigma-Aldrich
1-Dodecanol, ACS reagent, ≥98.0%