Merck
  • Prospective randomized comparison of oral sodium phosphate and sennoside A+B calcium lavage for colonoscopy preparation.
Prospective randomized comparison of oral sodium phosphate and sennoside A+B calcium lavage for colonoscopy preparation.
Surgical laparoscopy, endoscopy & percutaneous techniques (2011-04-08)
Manuk N Manukyan, Kerem Tolan, Utku Severge, Wafi Attaallah, Abut Kebudi, Asim Cingi
ABSTRACT
The aim of this study is to assess 2 different bowel-cleansing agents. The patients were prospectively randomized to 2 arms of sodium phosphate versus Sennoside A+B calcium preparation. Laboratory assessment, body weight, height, and vital signs were obtained at baseline and before colonoscopy. A self-administered questionnaire was completed by the patients. The time taken to complete the colonoscopy and the segment of the colon examined were recorded. The patients in the Sennoside A+B calcium group were more comfortable with the taste of the solution. Patients using sodium phosphate faced more nausea and significantly lower Ca levels and P values. The pulse rate was significantly higher in this group. Patients in the sennoside group had better grades of bowel cleansing in sigmoid and descending segments of the colon. Sennoside A+B calcium is more effective in some of the colonic segmental cleansing, causes fewer changes on serum electrolyte levels, and is better tolerated.
Product Description
Supelco
Sennoside A, analytical standard
Sennoside B, primary reference standard