Merck
  • Highly sensitive GC/MS/MS method for quantitation of amino and nonamino organic acids.
Highly sensitive GC/MS/MS method for quantitation of amino and nonamino organic acids.
Analytical chemistry (2011-03-11)
Hans F N Kvitvang, Trygve Andreassen, Tomas Adam, Silas G Villas-Bôas, Per Bruheim
ABSTRACT
Metabolite profiling methods are important tools for measurement of metabolite pools in biological systems. While most metabolite profiling methods report relative intensities or depend on a few internal standards representing all metabolites, the ultimate requirement for a quantitative description of the metabolite pool in biological cells and fluids is absolute concentration determination. We report here a high-throughput and sensitive gas chromatography/tandem mass spectrometry (GC/MS/MS) targeted metabolite profiling method enabling absolute quantification of all detected metabolites. The method is based on methyl chloroformate derivatization and quantification by spiking samples with metabolite standards separately derivatized with deuterated derivatization reagents. The traditional electron impact ionization is replaced with positive chemical ionization since the latter to a much larger extent preserve the molecular ion and other high molecular weight fragments. This made it easier to select unique MS/MS transitions among the many coeluting metabolites. Currently, the novel GC/MS/MS method comprises 67 common primary metabolites of which most belong to the groups of amino and nonamino organic acids. We show the applicability of the method on urine and serum samples. The method is a significant improvement of present methodology for quantitative GC/MS metabolite profiling of amino acids and nonamino organic acids.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
β-Alanine, 99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
β-Alanine, BioXtra, ≥99.0% (NT)
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
β-Alanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
N-Acetyl-L-glutamic acid, ReagentPlus®, 99%
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Sigma-Aldrich
5-Aminovaleric acid, 97%