- Charge mapping in 3(10)-helical peptide chains by oxidation of the terminal ferrocenyl group.
Charge mapping in 3(10)-helical peptide chains by oxidation of the terminal ferrocenyl group.
Organic letters (2011-02-24)
Alessandro Donoli, Vanessa Marcuzzo, Alessandro Moretto, Claudio Toniolo, Roberta Cardena, Annalisa Bisello, Saverio Santi
PMID21341757
ABSTRACT
Two series of 3(10)-helical peptides of different lengths and rigidity, based on the strongly foldameric α-aminoisobutyric acid and containing a terminal ferrocenyl unit, have been synthesized. Oxidation-state sensitive spectroscopic tags of helical peptides, the N-H groups, allowed mapping of the charge delocalization triggered by oxidation of the terminal ferrocenyl moiety and were monitored by IR spectroelectrochemistry.
MATERIALS